Deflected Point Septum Penetration Needles, Cadence Science®
Fournisseur: Cadence Science
Certificats
A propos de cet article
The deflected needle features point positioning on the axis of the needle lumen
This improved point geometry provides easier penetration and minimizes coring and tracking (particularly with finer gauge needles)
Ordering information: Non-sterile. Packaged 12 needles per box.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.