O-Ring Kit, Buna-N, Ace Glass Incorporated
# de catalogue 89235-688
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Buna-N O-Ring Kit
- Cat. no.:89235-688
- Supplier No.:7855-499
Spécifications
A propos de cet article
This Buna-N o-ring kit is packaged in a clear, plastic box with a re-sealable lid that lets you see the o-rings inside
Buna-N is also known as Nitrile rubber, is a synthetic blended rubber, co-polymer of acrylonitrile and butadiene. Generally resistant to fuels and oils. Generally black or brown in color.
It opens to 30 compartments, each clearly marked with the rubber type, o-ring size, and quantity included. The cardboard insert indicates the codes for each size, so you can easily re-order the o-rings individually as they are depleted. The quantity of each o-ring varies by size and compartment, but the kit contains 500 total o-rings.
Ordering information: The following sizes are included: -006, -007, -008, -009, -010, -011, -012, -110, -111, -112, -113, -114, -115, -116, -210, -211, -212, -213, -214, -215, -216, -217, -218, -219, -220, -221, -222, -325, -326, and -327.
Customers Who Bought This Also Bought