O-Rings, Buna-N, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Buna-N o-rings
Buna-N also known as Nitrile rubber, is a synthetic blended rubber, co-polymer of acrylonitrile and butadiene. It is generally resistant to fuels and oils, and is usually black or brown in color.
Frequently Bought Together