O-Rings, Viton, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Viton o-rings
Viton is a class of elastomers comprised of linear co-polymer of vinylidine flouride and hexafluoropropylene. Fluorine content is between 66 to 70%. Generally black in color, viton o-rings provide generally good temperature and chemical resistance.
Customers Who Bought This Also Bought