FHP NanoTech Microfiber Cloth, Vileda Professional-FHP
Supplier: Avantor
Certificates
About this item
NanoTech Micro is the solution for all operations and environments where maximum touch area hygiene is paramount.
- Nano silver particles are embedded in the endless microfibers
- Proven cleaning efficacy
- No growth of bacterial odors or germs on cloths for more than 24 hours
- Extremely low linting cloths
- Omit tumble drying, as cloths can be stored in moist conditions for 24 hours without bacteria odor growth
- Maintains antibacterial properties after 300+ laundry cycles
- Color-coded to prevent cross contamination
The NanoTech microfiber cloth's streak-free performance provides one-step cleaning with no drying cycle. The cloths can be used pre-prepared or via spray bottle method, giving users a variety of cleaning methods. Different color options reduce cross contamination. Reduce cost and eliminate odor with minimal chemical use.
Customers Who Bought This Also Bought