Syringe, Chromatography, LC Injection, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This syringe has an epoxy cemented 304 stainless steel needle permanently attached
Needles are 50mm long with 20° bevel tip.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.