Schwartz Drying Tubes, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Length indicated below is measured fromthe top of joint to the bottom of curve.
Tube O.D. is 17 mm, and included stoppers are [ST] 14/35. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought