Specifications
- Description:10-Layer CellCube® Module with 8,500 cm2 Growth Surface
- Material:Polycarbonate/Polystyrene
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Media volume:600 ml
- Surface:Tissue Culture Treated
- Cat. no.:89090-252
- Growth Area:8,500 cm2
- Sterility:Sterile
- Supplier No.:3200
Specifications
About this item
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment
It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth. These CellCube Modules provide: a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates, 8,500cm2 polystyrene growth surface optimally treated for the growth of attachment dependent cells, a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen, an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. Square Flask Shape.