Vacuum Plate and Pad
Fournisseur: VWR International
Certificats
A propos de cet article
This vacuum plate and pad are the perfect tools for vacuum experiments.
Here’s the perfect plate for your experiments requiring a vacuum situation. It works with bell jars and a vacuum pump – plus, no messy vacuum wax is needed. This handy plate for any lab features a 50 mm center hole, safety screw plug, and stopcock with hose nipple for vacuum tubing of 1/4" ID.
- No Messy Vacuum Wax Needed
- Air-Tight Contact
- Ideal with Bell Jars and Vacuum Pumps
- Center Hole: 50 mm
- Safety Screw Plug
They work with bell jars and vacuum pumps, and no messy vacuum wax is needed. The handy plate features a 50 mm center hole, safety screw plug, and stopcock with hose nipple for vacuum tubing of 1/4" ID.
The top plate measures 11" in diameter. It's made of cast iron.
Customers Who Bought This Also Bought