Immunotechnology Activity Kit
# de catalogue 470150-030
Fournisseur: G-Biosciences
Spécifications
- Description:Immunotechnology Activity Kit, Immunoprecipitation Technique
- Cat. no.:470150-030
- Supplier No.:BE-506
Spécifications
A propos de cet article
Hands-on Study of Immunoprecipitation
This kit contains all the reagents necessary to immunoprecipitate a specific protein from a complex sample. This hands-on activity focusing on immunoprecipitation permits the purification of specific proteins for which an antibody has been raised. The kit includes enough materials for six groups of four students to complete the experiment.
This kit contains all the reagents necessary to immunoprecipitate a specific protein from a complex sample. This hands-on activity focusing on immunoprecipitation permits the purification of specific proteins for which an antibody has been raised. The kit includes enough materials for six groups of four students to complete the experiment.
Customers Who Bought This Also Bought