ContactUsLinkComponent
Ward's® AP Biology Investigation 9: Genetics and Information Transfer: Restriction Enzymes
Ward's® AP Biology Investigation 9: Genetics and Information Transfer: Restriction Enzymes
# de catalogue 470134-776
Fournisseur:  Avantor
RESTRICTION ENZYMES AP BIO. INVEST. 9
Ward's® AP Biology Investigation 9: Genetics and Information Transfer: Restriction Enzymes
# de catalogue 470134-776
Fournisseur:  Avantor
Spécifications
  • Description:
    AP Investigation 9: Restriction Enzymes Kit
  • No. of Lab Groups:
    8
  • Cat. no.:
    470134-776
Spécifications
A propos de cet article
Meets AP Science Practices 3 and 6
The Only Kits Aligned with AP Science Practices
Ward's AP Biology Investigations provide complete coverage of the Four Big Ideas and Seven Science Practices to help you meet new AP Biology curriculum standards and prepare students for real-world success in college and research-level labs.
This complete series of investigations emphasizes the progression to fully student-directed investigations and builds competence in all of the science practices, helping students:
•Practice concept application instead of fact memorization.
•Focus on self-guided experimentation instead of step-by-step lab procedure.
•Investigate real-world concepts with greater curriculum flexibility.
•Apply problem-solving skills to strengthen student inquiry.
Each investigation follows a series of three parts, beginning with a structured lab investigation, followed by a related guided inquiry investigation with thoughtful assessment questions, and finally, suggestions to encourage students to formulate their own questions and explore further self-guided experimentation.
In this investigation...
In this lab, students will use electrophoresis technology to analyze DNA sequence by fragment analysis. Simulating the results of a Southern Blot DNA Fingerprint, students will compare fragments of known size to fragments of an unknown size to calculate relative molecular weights and match fragment patterns. Further self-guided experimentation is explored as they investigate parameters of various factors that relate to DNA technology and restriction enzyme analysis. Investigation includes updated literature aligned with new AP Biology curriculum standards for 2012. Meets AP Science Practices 3 and 6, and Big Idea 3. Materials provided for 32 students in eight lab groups.
Note: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.
  • Solve a crime using real- life forensic techmiques
  • Model DNA structure and restriction enzyme activity
In this investigation, students will use electrophoresis technology to analyze DNA sequence by fragment analysis. Simulating the results of a Southern Blot DNA Fingerprint, students will compare fragments of known size to fragments of an unknown size to calculate relative molecular weights and match fragment patterns. Further self-guided experimentation is explored as they investigate parameters of various factors that relate to DNA technology and restriction enzyme analysis.
The Only Kits Aligned with AP Science Practices. Ward's AP Biology Investigations provide complete coverage of the Four Big Ideas and Seven Science Practices to help you meet new AP Biology curriculum standards and prepare students for real-world success in college and research-level labs. Each investigation follows a series of three parts, beginning with a structured lab investigation, followed by a related guided inquiry investigation with thoughtful assessment questions, and finally, suggestions to encourage students to formulate their own questions and explore further self-guided experimentation. This complete series of investigations emphasizes the progression to fully student-directed investigations and builds competence in all of the science practices, helping students:
• Practice concept application instead of fact memorization.
• Focus on self-guided experimentation instead of step-by-step lab procedure.
• Investigate real-world concepts with greater curriculum flexibility.
• Apply problem-solving skills to strengthen student inquiry.
AP and Advanced Placement Program are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
BodyTextAccessories: Refill kits contain only consumable component items and do not contain additional copy masters.
Ordering information: Included coupon can be redeemed by mail, phone, fax, or email.
Kit contains the following materials for 8 groups of students: coupon for perishable materials, felt tip markers, prepared agarose, staining tray, pop beads with connectors, microcentrifuge tubes, buffer, instructions, and copy masters.
Customers Who Bought This Also Bought