Test Tubes, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Test tubes, available with or without matching glass stoppers; [ST] 24/40 stopper is hollow; #16 and #19 stoppers are solid.
Fabricated from borosilicate glass.
Ordering information: Available complete with stopper and tube, or components can be purchased separately.
Customers Who Bought This Also Bought