A propos de cet article
IBI's Luria Broth (LB Miller) is used in molecular genetic studies.
- Ensure highest lot-to-lot quality and consistency
- Maintainiand propagate E. coli in molecular and microbiology procedures
- Sodium chloride provides essential electrolytes
IBI growth media products start with raw materials sourced only from North America and European suppliers, ensuring the highest lot-to-lot quality and consistency
IBI's LB Miller broth contains Casein Digest Peptone (10 g/L), Sodium Chloride (10 g/L), and Yeast Extract (5 g/L) with a pH (@25.0°C) of 7.0 ± 0.2. To prepare, mix 25.0 gm of the medium in 1L of purified water until evenly dispersed. Heat with repeated stirring and boil for 1 minute to dissolve completely. Distribute and autoclave at 121.0 °C for 15 minutes.
Store at room temperature, keep tightly sealed, and protect from moisture and light.
IBI's Luria Broth (LB Miller) is used in molecular genetic studies, as well as maintaining and propagating E. coli in molecular and microbiology procedures. The inclusion of casein peptone and yeast extract supply essential growth factors; such as nitrogen, sulfur, minerals, and vitamins. Sodium chloride provides essential electrolytes.