MagLab™ Lab Storage Series
Fournisseur: Heathrow Scientific
Certificats
A propos de cet article
Easily organize common lab products off your benchtop with the MagLab storage system.
- Made of robust ABS plastic in bright translucent colors
- Secure direct to metal with built-in powerful magnets or non-metal surfaces with included self-adhesive mounting plate
Create lab storage space-off your benchtop.
*Kimwipes is a registered trademark of Kimberly-Clark Corp.
Each rack is made of robust ABS plastic in a bright translucent color and features four powerful magnets for a secure hold onto most metal surfaces. MagRack™ holds four 50 ml tubes and nine 15mL tubes. MagWipe™ holds a standard (11×21cm) Kimwipes®* box. MagPette™ holds two pipettors (fits most popular styles). MagBox™ holds miscellaneous accessories. The MagLab Series is available individually or as a set of four.
Customers Who Bought This Also Bought