Support Stand, 'C' Shape, All Stainless Steel, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
An all-stainless-steel support stand with a heavy, six pound base
This stand can accommodate vessels up to 18" in diameter within its 'C' shape. The support rod is ⁵/₈" in diameter, fastened to the base via stainless steel lock nuts. There are two additional threaded holes in the base legs to accommodate extra support rods.
Ordering information: Complete item consists of one base and one rod. Additional support rods may be purchased separately.
Customers Who Bought This Also Bought