Impinger, Sherer, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This Sherer-type impinger consists of bottle with hex base and 45/50 top joint, and either an SO2 bubbler tube with 25 mm diameter Porosity C (25-50 micron) fritted disc, or an impinger stopper
Capacity- 275mL. Inlet/outlet tubes are 8mm. Fabricated from borosilicate glass.
Ordering information: Available as a complete item, or components may be purchased separately.
Customers Who Bought This Also Bought