Vapor Tubes, for Heidolph 4000 Series, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Used as replacements with Heidolph 4000 Series rotary evaporators.
- Fabricated from borosilicate glass
Tube is secured in rotary drive with low-stress plastic clip that seats into groove behind joint. Available plain or with Firestone "splash guard" to protect against splash-up.
Customers Who Bought This Also Bought