Connectors, with PTFE Plug Valve, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Polypropylene connectors, two-way or three-way, with PTFE valve and serrated tubulations
Takes 6.4 mm to 9.5 mm I.D. tubing.
Customers Who Bought This Also Bought