PTFE Stopcock Valve, Two Way, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This PTFE valve is used with NPT fittings.
- Various ends available
- Stopcocks and compression ferrules made of virgin PTFE
- Nuts and ferrule-gripper made of PVDF
Various ends available, including male and female NPT, luer, ¼" tube and ¼"-28 UNF. IPS PTFE Stopcocks are made of virgin PTFE. Compression ferrules are also made of virgin PTFE. Nuts and ferrule-gripper are made of PVDF.
Customers Who Bought This Also Bought