ContactUsLinkComponent
VWR® Thermal Shake Touch Thermoshaker with 1.5 ml Block
VWR® Thermal Shake Touch Thermoshaker with 1.5 ml Block
Fournisseur:  VWR International
Certificats
VWR® Thermal Shake Touch Thermoshaker with 1.5 ml Block
VWR® Thermal Shake Touch Thermoshaker with 1.5 ml Block
Fournisseur:  VWR International
Certificats
Spécifications
  • Catalog No:
  • 10027-174
  • 89232-908
  • Temperature accuracy:
  • ±1° between 20° to 45 °C, ±2° above 45 °C
  • ±1° between 20° to 45 °C, ±2° above 45 °C
  • Speed range:
  • 300 to 3000 RPM
  • 300 to 3000 RPM
  • Speed accuracy:
  • ±2 %
  • ±2 %
  • Overall dimensions:
  • 26L x 24.8W x 13.2H cm (10.25 x 9.75 x 5.2")
  • 26L x 24.8W x 13.2H cm (10.25 x 9.75 x 5.2")
  • Application:
  • Heating Only
  • Heating Only
  • Orbit:
  • 3 mm
  • 3 mm
  • Temperature range:
  • 4° above ambient to 100 °C
  • 4° above ambient to 100 °C
  • Unit weight:
  • 3.7 kg. (8 lbs.)
  • 3.7 kg. (8 lbs.)