Needles, Standard Hypodermic, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Standard hypodermic stainless steel needles with 12° regular medical point tip and female Luer-Lok hub
Packed 12 needles on card-cellophane wrapped. Each card is individually boxed.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.