Needles, Special Length, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Special length 304 stainless steel needles with a deflected septum point and standard female hub
The deflected septum point is equivalent to B-D Huber or Hamilton Style 1 and 2.
Packaging: Packed 6 per package or a case of 12.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.