Fixed Volume Microdispensers, Drummond Scientific
Fournisseur: Drummond
Certificats
A propos de cet article
Fixed Volume Microdispensers have an accuracy of ±1% at full volume.
- Borosilicate Glass Bores
- Stainless Steel Plungers
- Accuracy at Full Volume: ±1%
The microdispensers feature borosilicate glass bores with stainless steel plungers.
All plungers 25 µL or larger feature PTFE tips.
Customers Who Bought This Also Bought