Midget Impinger, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This piece is used for sampling small air volumes at a low jet velocity
Bottle has encircling graduations from 0 to 25 mL, or 0 to 30 mL in 5 mL divisions. Nozzle is calibrated to deliver 0.09 to 0.11 CFM at 30.5 cm (12-inch) H2O vacuum. Used in determination of oxidants: Alkaline Potassium Method. Joint is [ST] 24/40 outer, tooled with reinforcement rings for added strength and stability. Inlet/outlet are 7 mm O.D. Fabricated from borosilicate glass.
Ordering information: Complete item consists of bottle and tube. Items can also be purchased separately.
Customers Who Bought This Also Bought