Joint/Flask Adapters Without Combi Clip, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This is a replacement receiving flask adapter for Buchi rotary evaporators.
- For Buchi rotary evaporators
- Without combi clip
- Fabricated from borosilicate glass
Joint to flask adapter joins the steam tube to the receiving flask. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought