Cap, GL, with Hole, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This open top red polybutylene teraphthalate (PBT) cap is for use with GL threads
When used with a hose connection and appropriate sealing ring, this cap will accommodate tubing of the same approximate diameter. Temperature range is –45ºC to 180ºC.
Customers Who Bought This Also Bought