Midget Impinger, Modified, Plain Nozzle, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This modified midget impinger has a plain nozzle
Bottle has top [ST] 24/40 outer joint tooled with reinforcement rings for added strength and stability. Tube is modified with [SJ] 12/5 joints, both vertical (use 89060-252 pinch clamp). The out [SJ] Joint is 12/5 Ball. Bottle is graduated 0 to 25 mL in 5 mL divisions. Fabricated from borosilicate glass.
Ordering information: Complete item includes tube and 89059-294 bottle. May also be purchased separately.
Customers Who Bought This Also Bought