SP Bel-Art Test Tube Capper, Bel-Art Products, a part of SP
# de catalogue 89233-990
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:Test Tube Capper, 44×89 m m (1¾ × 3½”)
- Cat. no.:89233-990
- Supplier No.:F187990000
Spécifications
A propos de cet article
The vinyl-covered handhold fits comfortably in the palm of the hand while the polypropylene tip presses down on the cap to securely position it within the test tube.
- Vinyl-covered handhold
- Polypropylene tip secures cap in position
- Reduced finger soreness and hand stress
Capping can be performed on glass or plastic test tubes in a rack, or while being held in the hand
With the Scienceware® Test Tube Capper, you can use your entire hand for leverage, reducing finger soreness and hand stress.
It is designed for use with recessed center test tube caps.
Customers Who Bought This Also Bought