Adapter, Feed Tube, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This adapter is threaded at both ends and on side joint.
- Top and side nylon bushings and FETFE o-rings included
- One silicone rubber septa also included
- Fabricated from borosilicate glass
The #7 Ace-Thred joints at top and side are each supplied with a nylon bushing and FETFE o-ring. One silicone rubber septa is also included. Size listed also refers to the I.D. of the tubing above the lower thread, and this corresponds to the column diameter.
Customers Who Bought This Also Bought