Feed Tube with Hose Connection, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This feed tube permits packing of column without excessive drop impact
Used in conjunction with a nylon bushing. Supplied straight or bent. Hose connection size C for use with 5/16" or 3/8" I.D. tubing. Glass tube is 7 mm O.D. Length is 76 mm (3"). Fabricated from borosilicate glass.
Also used for side arm introduction of effluent, or may be used for pressurized operation.
Customers Who Bought This Also Bought