Feed Tube, Luer-Lok Tip, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This feed tube with Luer-Lok tip permits packing of column without excessive drop impact
- Glass tube is 7 mm O.D
- Fabricated from borosilicate glass
Also used for side arm introduction of effluent, or may be used for pressurized operation. Used in conjunction with a nylon bushing.
Customers Who Bought This Also Bought