ContactUsLinkComponent
Anaerobic Culture Tube, Chemglass Life Sciences
Anaerobic Culture Tube, Chemglass Life Sciences
# de catalogue 89167-178
Fournisseur:  Chemglass
Small Business Enterprise
Tube W/Stopper 18X150Mm
Anaerobic Culture Tube, Chemglass Life Sciences
# de catalogue 89167-178
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Ø:
    18 mm
  • Description:
    Anaerobic Culture Tube
  • Height:
    150 mm
  • Supplier No.:
    CLS-4209-01
Spécifications
A propos de cet article
COMPLETE anaerobic tubes, 18×150 mm, supplied with 20 mm blue butyl rubber stopper and aluminum seal.
Customers Who Bought This Also Bought