Specifications
- Light source:Tungsten-halogen
- Description:Spectronic 200 Visible Spectrophotometer
- Spectral bandwidth:≤ 4 nm
- Photometric range:-0.3 A to 2.5 A
- Optical design:Single Beam
- Stray light:< 0.2 %T (with SPECTRONIC standard SRM 400 filter)
- Photometric accuracy:± 0.01 A at 0.3 A
- Electrical:100 –240 V, 50 –60 Hz (selected automatically)
- Wavelength range:340 - 1000 nm
- W×D×H:39×30×16 mm (15.3×11.8×3.6")
- Cat. no.:97054-250
- Supplier No.:222-265700
Specifications
About this item
SPECTRONIC 200 is a modern, visible spectrophotometer for education and routine industrial use
USB connectivity provides additional flexibility for data and method storage, printing, or connection to a PC with VISIONlite 5 (or later) software for more involved experiments, data processing and storage.
Built upon a foundation of over 50 years of experience the SPECTRONIC 200 features everything your existing visible spectrophotometer offers and more. Familiar controls, a full color display, and built-in software provides a full range of functionality including live display mode, full spectrum measurements, multi-wavelength analysis, and quantitative analysis with up to 4 standards. A programmable analyzer mode allows you to streamline repetitive procedures.