Coupling, Michel-Miller, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These precision made PTFE couplings have a 316 stainless steel liner for connecting Michel-Miller and other columns when packing.
Designed to make a leak-tight seal without the use of o-rings. Simply tighten until white ring appears indicating a seal has been made, and secure lock nut. No need to tighten further. Male thread is size designation, and matches thread size of columns.
Customers Who Bought This Also Bought