Adapter, End Fitting, Michel-Miller, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This is a precision-made Michel-Miller replacement PTFE end fitting adapter.
- Leak-tight seal without o-rings
- Male thread is size designation matching column
- Top female thread is 1/4-inch-28 UNF
- Three options for retaining packing materials
Adapter is designed to make a leak-tight seal without the use of o-rings
There are three different options used to retain packing material: 1) paper filter discs; Porosity D (10-20 micron) glass filter discs; or PTFE screens. Choose the option that best meets your needs.
Simply tighten until white ring appears indicating a seal has been made, then secure lock nut; no need to over tighten. Male thread is size designation and matches thread size of column. Female thread at top is 1/4-inch-28 UNF with Heli-Coil insert to accept flanged end tubing fittings. Bore is 1.5mm (.060-inches).
Ordering information: Glass filter disc is supplied with appropriate adapters; adapters using paper filter discs or PTFE screens do NOT include those items. To retain packing material: Paper filter discs 89052-068, 89052-496, 89052-076, 89052-078, 89052-080; Porosity D (10-20 micron; 89052-860) glass filter discs 89052-082, 89052-084, 89052-086 or PTFE screens 89052-498, 89059-192.
Customers Who Bought This Also Bought