Stopper, Michel-Miller, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These precision made PTFE stoppers are for use at either end of Michel-Miller columns, or on other Ace-Thred connections, when a leak-tight seal without the use of o-rings is desired
Customers Who Bought This Also Bought