Adapter, Column, Feed Tube, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This adapter is threaded at both ends.
- Top is #7 Ace-Thred
- Lower thread
- Fabricated from borosilicate glass
The top of this adapter has a #7 Ace-Thred that is supplied with a nylon bushing and FETFE o-ring for use with adjustable feed tubes. Size listed also refers to the I.D. of the tubing above the lower thread, and this corresponds to the column diameter.
Ordering information: For extra bushings (includes two FETFE o-rings), order 89051-122. For o-ring only, order 89060-958 (package of 12).
Customers Who Bought This Also Bought