Preparative Funnel, Chromatography, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Bulb type reservoir preparative funnel for chromatography preparative work.
- Both ends have Ace-Thred connections
- Top can be sealed with a solid plug
- Fabricated from borosilicate glass
Customers Who Bought This Also Bought