Filter Column, Michel-Miller, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These columns are for use with the Michel-Miller liquid chromatography System as a filter to assure only clean solvent enters column.
Threads are #7 Ace-Thred for use with 89052-068 end fitting. Usually positioned, in line, between the pump and the column using 1/4-inch-28 UNF tubing connectors. Fabricated from borosilicate glass.
Ordering information: Glass only is supplied. Fittings must be ordered separately. Filter material, such as glass wool, not supplied.
Customers Who Bought This Also Bought