Adapter, Michel-Miller, Injection Port, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
A simple, all-PTFE septum injector that allows for the most efficient use of Michel-Miller non-packed columns.
- Direct on-column injection
- Leak-tight seal without o-rings
- Threaded
Injector allows direct on-column injection to the center of the filter disc at top of column without stopping the solvent flow. Designed to make a leak-tight seal without the use of o-rings. Simply tighten until white ring appears indicating a seal has been made, then secure locknut; no need to over tighten. Top thread is 5/16-inch-24 UNF for use with injection port cap that holds a septum. Male thread is size designation and matches thread size of columns. Bore is 1.5mm (.060-inches).
Ordering information: Adapter is available with or without Heli-Coil insert. Supplied with one PTFE coated silicone rubber septum. Use paper filter discs (not included) to retain packing material. Not suitable for larger chromatography columns. Contact your Avantor representative for details.
Customers Who Bought This Also Bought