ContactUsLinkComponent
Syringe, Gas Tight, Luer-Lok, Ace Glass Incorporated
Syringe, Gas Tight, Luer-Lok, Ace Glass Incorporated
Catalog # 89053-708
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Syringe, Gas Tight, Luer-Lok, Ace Glass Incorporated
Catalog # 89053-708
Supplier:  Ace Glass
Supplier Number:  5932-08
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Description:
    Luer-Lok Gas Tight Syringe
  • Cat. no.:
    89053-708
  • Supplier No.:
    5932-08
Specifications
About this item
Syringe with PTFE coated plunger and Luer-Lok
Used for sample introduction in chromatography solvent delivery systems. Has a capacity of 5mL.