Axygen® Research-Grade Bevelled Reference Pipette Tips 200 µl, Corning
Supplier: Corning
Certificates
About this item
Axygen tips are designed to fit a wide variety of single-channel and multichannel pipettors.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry.
Specifications
- Catalog No:
- 47734-742
- 10011-376
- 10011-378
- 95042-896
- 10011-894
- 10011-934
- 47734-722
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa