Axygen® Pipette Filter Tip, Axygen®, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Axygen brand filter tips allow free passage of air while preventing passage of aerosols.
The filters contain no leachable additives.
Customers Who Bought This Also Bought