PCR Plates, Sorenson BioScience
Fournisseur: Sorenson Bioscience
Certificats
A propos de cet article
UltraAmp 96- and 384-well PCR plates contains bold-printed alphanumerics and fit all leading thermal cyclers, DNA sequencing/detection instruments, and Real-Time PCR instruments.
- Tight manufacturing tolerances and SBS standard dimensions create plates ideal for automated processes
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
Customers Who Bought This Also Bought