PCR Tube and Domed Cap Strips, 0.2 ml, Sorenson BioScience
Supplier: Sorenson Bioscience
Certificates
About this item
PCR tube and cap strips are manufactured from medical grade polypropylene in a tightly controlled production environment and feature a think-wall design for optimal heat transfer.
- Tubes fit most popular thermal cyclers
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
Customers Who Bought This Also Bought