Barrier Pipette Tips, Sorenson BioScience
Supplier: Sorenson Bioscience
Certificates
About this item
The barrier pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment and feature an inert hydrophobic aerosol-blocking barrier.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips minimize sample retention.
Customers Who Bought This Also Bought