PCR Plate Sealers, Sorenson BioScience
Supplier: Sorenson Bioscience
Certificates
About this item
Adhesive seals and cap strips prevent evaporation during thermal cycling or storage and are compatible with Sorenson BioScience PCR plates and tube strips.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
Frequently Bought Together