WHEATON® Peristaltic Pumps, Dispensing Pumps and Tubing
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Wheaton peristaltic pumps are used for dispensing and general liquid handling purposes
The pumps are delivered ready to use with tubing kits included. The pumps can be programmed to dispense measured aliquots into culture plates, vials, tubes or bottles. The Omnispense units have additional features including speed ramping and totalizer/flow mode.
Ordering information: Includes complete pump, instruction manual and tubing kits. Works with 2.3mm thick tubing. Additional pump heads and foot switches available.
Spécifications
- Catalog No:
- 25485-031
- 16171-481
- 25485-037
- 25485-029
- 25485-030
- 89194-340
- 25485-035
- 25485-038
- 89194-348
- 89194-346
- 89194-344
- 89194-342
- 89194-356
- 89194-354
- 89194-352
- 14231-318
- 89194-350
- 14230-632
- 89194-338
- 89194-336
- 14231-588
- 89212-192
- 89212-184
- 89212-186
- 14231-582
- 89212-188
- 14231-594
- 89212-190
- 89194-334
- 89194-332
- 14230-630
- 89194-330
- 89212-182
- 89212-194
- 89212-196
- 14006-832
- 89212-198
- 14006-836
- 89212-200
- 14006-834
- 14006-838
- Model:
- UniSpense PRO
- UniSpense PRO
- UniSpense PRO
- UniSpense PRO
- UniSpense PRO
- UniSpense PRO
- UniSpense PRO
- OmniSpense ELITE
- OmniSpense ELITE
- OmniSpense ELITE
- Min. flow rate:
- 20.4 ml/min
- 30 ml/min
- 30 ml/min
- 30 ml/min
- 20.4 ml/min
- 30 ml/min
- 30 ml/min
- 30 ml/min
- 30 ml/min
- 20.4 ml/min
- 20.4 ml/min
- 20.4 ml/min
- 20.4 ml/min
- 20.4 ml/min
- Max. flow rate:
- 1746 ml/min
- 1080 ml/min
- 1080 ml/min
- 1080 ml/min
- 1746 ml/min
- 1080 ml/min
- 1080 ml/min
- 1080 ml/min
- 1080 ml/min
- 1746 ml/min
- 1746 ml/min
- 1746 ml/min
- 1746 ml/min
- 1746 ml/min