Axygen® Automation Tips for Qiagen BioRobot® Series, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Axygen® offers a comprehensive line of tips for Qiagen® automation systems.
- Black tips are conductive for liquid level sensing protocols
- Optimised fine point for accurate dispensing
- Certified RNase-, DNase- and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
Spécifications
- Catalog No:
- 89124-962
- 89124-978
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa