Axygen® Robotic Pipette Tips, PK Series, Corning
Fournisseur: Corning
Certificats
A propos de cet article
The PK series robotic tips for the PerkinElmer® and Packard® Systems are certified RNase-, DNase-, and pyrogen-free.
The PK series robotic tips are engineered to exact dimensions of OEM tips to ensure automation compatibility.
Spécifications
- Catalog No:
- 80086-040
- 80086-038
- 80086-042
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa